Ultimono ; Performance degraded ; low beam current
Issue
Ultimono performance degraded and screen current very low
Environment
Spectra 200, Spectra 300, Spectra C, Spectra Ultra
Resolution
These are possible resolutions for the issue:
- Perform Ultimono alignment procedure (4022 190 63889 )
Cause
These are possible causes for the issue:
- Wrong ultimono factor in Feg register
Details
After loading Factory Ultimono alg file and Feg register the optimal Ultimono value changed.
This resulted in suboptimal conditions to perform Ultimono energy resolution and to obtain a sufficient beam current for the energy resolution.
Attachment(s)
File | Last Modified |
---|---|
clipboard_ea16aa5ae36a092029a49941a7fabd14c.png | June 16, 2022 |
clipboard_e166ef303add7cb4721d9ece1e2df25dc.png | June 16, 2022 |