Workset menu is in the middle of the screen / Workset position
Issue
Incorrect Workset position
Environment
Titan Krios G3
Resolution
These are possible resolutions for the issue:
- Disable nView
Cause
These are possible causes for the issue:
- Nvidia desktop manager (nView) is enabled
Details
Workset menu is in the middle of the screen instead of top left corner.
Solution : Right click on desktop, select nView Desktop Manager click on disable and restart TEM User interface.
Make sure that nView is disabled in both Customerservice and Supervisor account.
Attachment(s)
File | Last Modified |
---|---|
clipboard_ea6d36a7a273a63f9a1f4d094759b45cd.png | June 16, 2022 |
clipboard_e781a94c28f5c602b8cbf38a241c3e3fd.png | June 16, 2022 |
clipboard_e88cc499d3c77cf70b3178b986a4c7050.png | June 16, 2022 |