1507060 - Rev 3.2 - Multidrop DW - User Manual
The Thermo Scientific Multidrop DW User Manual provides comprehensive guidance for the operation, installation, and maintenance of the instrument. The instrument can dispense up to eight different reagents into deep-well plates, 96-well plates, and tube racks. Features include high-speed dispensing, superior accuracy, and an autoclavable dispensing cassette.
Description:
User manual
Environment:
Thermo Scientific Multidrop® DW
Attachment(s):
| File | Last Modified |
|---|---|
| 1507060 - Rev 3.2 - Multidrop DW - User Manual.pdf | December 01, 2021 |