7011355 - 1300 Series A2 Class II Type A2 Biological Safety Cabinet - Operating Manual
The document titled "7011355 - 1300 Series A2 Class II Type A2 Biological Safety Cabinet - Operating Manual" provides comprehensive operational instructions for the 1300 Series A2 Class II Type A2 Biological Safety Cabinets. It includes guidelines on safe usage, maintenance procedures, troubleshooting tips, and technical specifications. The manual ensures that users can effectively and safely operate the safety cabinet, maintaining its performance and adhering to safety standards.
Description:
Operating manual, 2007 - 2010. Revisions 0, 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, 16, 17, 18, 19, and 20.
Environment:
Thermo Scientific 1300 Series Class II, Type A2 biological safety cabinet models:
| Stainless Steel Interior | Painted Interior | Size | Voltage | ||
| 10 inch opening | 8 inch opening | 10 inch opening | 8 inch opening | ||
| 1354 | 4ft | 230V, China | |||
| 1345 | 1305 | 1355 | 1325 | 4ft | 120V |
| 1346 | 1306 | 1356 | 1326 | 4ft | 230V |
| 1347 | 1307 | 1357 | 1327 | 6ft | 120V |
| 1348 | 1308 | 1358 | 1328 | 6ft | 230V |
| 1359 | 6ft | 230V, China | |||
| 1384* | 4ft | 230V, China | |||
| 1375 | 1335 | 1385* | 1395 | 4ft | 120V |
| 1376 | 1336 | 1386* | 1396 | 4ft | 230V |
| 1377 | 1337 | 1387* | 1397 | 6ft | 120V |
| 1378 | 1338 | 1388* | 1398 | 6ft | 230V |
| 1389* | 6ft | 230V, China | |||
* Includes cabinet, adjustable height stand, UV light and armrest.