3100 Series - Resetting the Replace Hepa Filter Reminder
The purpose of this document is to guide users on resetting the Replace HEPA Filter reminder for the 3100 Series Model 3110. It includes steps to reset the timer and clear the REPLACE HEPA alarm, as well as instructions to set the reminder for a specified number of months (1 to 12 months) using the Mode key, right arrow, and Enter button.
Description:
Resetting the replace Hepa filter reminder
Environment:
3100 Series Model 3110
Attachment(s):
| File | Last Modified |
|---|---|
| 3100 Series - Resetting the Replace Hepa Filter Reminder.pdf | December 27, 2021 |