7190512 - Rev 7 - Qualified Installation of the Analog Board
The document "7190512 - Rev 7 - Qualified Installation of the Analog Board" provides instructions and guidelines for the qualified installation of the analog board in various series incubators. The relevant models include 310, 370, 3110, 3307, and 3950.
Description:
Qualified Installation of the Analog Board
Environment:
Series Incubators, models:
- 310
- 370
- 3110
- 3307
- 3950
| Table 1. Analog Output Board Specifications | |
| 1900044 (4-20 mA) - Output Scaling | |
| Temp | 0-100°C (12.5-162.5°C 370 and 3307 Series) |
| RH | 0-100 %RH |
| CO2 | 0.0-100.0 %CO2 |
| O2 | 0.0-100.0 %O2 |
| 1900045 (0-5V) - Output Scaling | |
| Temp | 0-100°C (0-150°C 370 and 3307 Series) |
| RH | 0-100 %RH |
| CO2 | 0-100.0 %CO2 |
| O2 | 0-100.0 %O2 |
| 1900046 (0-1V) - Output Scaling | |
| Temp | 0-100°C (0-150°C 370 and 3307 Series) |
| RH | 0-100 %RH |
| CO2 | 0-100.0 %CO2 |
| O2 | 0-100.0 %O2 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7190512 - Rev 7 - Qualified Installation of the Analog Board.pdf | December 21, 2021 |