Heracell 150i and 240i Temp High alarm
The document "Heracell 150i and 240i Temp High alarm" explains that the red frame around the temperature measurement signifies that the control system has detected the measured temperature exceeding the setpoint by more than 1°C in HERAcell 150i and 240i incubators.
Description:
The red frame around the temperature measurement indicates the controls has witnessed the measured temperature more than 1C above the Setpoint.
Environment:
150i
240i
Attachment(s):
| File | Last Modified |
|---|---|
| Temp High Red Frame on Heracell 150i and 240i.pdf | December 17, 2024 |