Reversing the door swing on a 160i Co2 Incubator
Provides guidance from the Vios 160i service manual to reverse the door swings (hinge side) on the 160i CO2 incubator. Summarizes the resolution: door can be reoriented by swapping hinges and latch, verifying gasket alignment, and confirming proper closure.
Description:
Instructions from service manual to reverse the door swing on Vios 160i. Also known as Reversing the door hinges.
Environment:
Vios 160i
160i
Attachment(s):
| File | Last Modified |
|---|---|
| Vios 160i Door Swing Reversal from Service Manual.pdf | December 08, 2025 |