What does the service interval mean
The document explains that the service interval in a CO2 incubator is a timed reminder for annual maintenance. This maintenance includes inspecting the door gasket, replacing the inline CO2 filter, and verifying the unit's calibration.
Issue
What does the service interval mean?
Environment
CO2 Incubator
Resolution
The service interval is a timed reminder to perform the annual maintenance, which includes inspecting the door gasket, replacing the inline CO2 filter, and verifying the calibration of the unit.