102219021 - Rev B - Napco Analog Display Automatic CO2 Water-Jacketed Incubator - Operating Manual
The document is the operating manual for the Napco analog display automatic CO2 water-jacketed incubators, covering models 5100, 5200, and 5300. It provides detailed instructions on the setup, operation, and maintenance of these incubators, including specifications for electrical service requirements.
Description:
Operating manual
Environment:
Napco analog display automatic CO2 water-jacketed incubator, model numbers:
| Model | Electrical Service | |||
|
525 Watts, 120 VAC 50/60 Hz, 4.4 Amps | |||
|
1000 Watts, 120 VAC 50/60 Hz, 8.3 Amps | |||
|
1000 Watts, 120 VAC 50/60 Hz, 8.3 Amps | |||
Attachment(s):
| File | Last Modified |
|---|---|
| 102219021 - Rev B - Napco Analog Display Automatic CO2 Water-Jacketed Incubator - Operating Manual.pdf | December 27, 2021 |