Thermo Fisher Scientific
50142486 - Rev 00 - VWR Collection Model 89511-416, Incubator Cooled 0.65CF - User Manual
Description: 
User manual
Environment:
VWR Collection Model 89511-416, Incubator Cooled 0.65CF, technical data:
Designation Value
Volume 18 liter
Dimensions (W x H x D) 10.24 x 16.34 x 18.50 in
Interior dimensions 7.87 x 12.20 x 12.20 in
Weight approx. 15.87 lb
Minimal wall distance 1.97 in on the sides and back
Number of stiffening plates Standard 2, maximum 3
Current consumption 0.45 Amps
Protection/Installation class II
Degree of protection IP20
Max. mechanical load 4.41 lb per plate, 11.02 lb per device
Connection voltage 100-240V AC +/-10% 50/60 Hz, Connection with the enclosed mains adapter