50142486 - Rev 00 - VWR Collection Model 89511-416 Incubator Cooled 0.65CF - User Manual
The document titled "50142486 - Rev 00 - VWR Collection Model 89511-416, Incubator Cooled 0.65CF - User Manual" provides user instructions for the VWR Collection Model 89511-416, Incubator Cooled 0.65CF. It includes technical specifications such as volume (18 liters), dimensions, weight, minimal wall distance, number of stiffening plates, current consumption, protection class, degree of protection, mechanical load capacity, and connection voltage. This manual is essential for proper setup, opera
Description:
User manual
Environment:
VWR Collection Model 89511-416, Incubator Cooled 0.65CF, technical data:
| Designation | Value |
| Volume | 18 liter |
| Dimensions (W x H x D) | 10.24 x 16.34 x 18.50 in |
| Interior dimensions | 7.87 x 12.20 x 12.20 in |
| Weight | approx. 15.87 lb |
| Minimal wall distance | 1.97 in on the sides and back |
| Number of stiffening plates | Standard 2, maximum 3 |
| Current consumption | 0.45 Amps |
| Protection/Installation class | II |
| Degree of protection | IP20 |
| Max. mechanical load | 4.41 lb per plate, 11.02 lb per device |
| Connection voltage | 100-240V AC +/-10% 50/60 Hz, Connection with the enclosed mains adapter |
Attachment(s):
| File | Last Modified |
|---|---|
| 50142486 - Rev 00 - VWR Collection Model 89511-416, Incubator Cooled 0.65CF - User Manual.pdf | December 22, 2021 |