Thermo Fisher Scientific
50151882 - Rev 00 - Isotemp Peltier Low Temperature Incubators - User Manual
Description: 
User manual for Fisherbrand™ Isotemp™ Peltier Refrigerated Incubator
Environment:
  • Isotemp Peltier Low Temperature Incubator 178L 120V Model Number 51031568
  • Isotemp Peltier Low Temperature Incubator 381L 120V: Model Number 51031569
Models   Isotemp 178L Isotemp 381L
Parameter Unit    
Chamber volume L-cu ft 178-6.3 381-13.5
Chamber temperature range °C-°F 10-60, 50-140