50153099 - Fisherbrand C02 Incubators - Operations Manual
The purpose of the operations manual for Fisherbrand CO2 Incubators is to provide detailed instructions and guidelines on the usage, maintenance, and troubleshooting of various models of Fisherbrand Isotemp CO2 incubators. The manual covers different models, including those with thermal conductivity (TC) and infrared (IR) CO2 sensor technologies, and provides information about their features such as the 90°C decontamination cycle and stainless steel 150L chamber.
Description:
Operations manual
Environment:
Fisherbrand C02 Incubators, models:
| Model | Product | Cat. No. |
| 50153428 | Fisherbrand Isotemp CO2 incubator, stainless steel 150L chamber with on-demand 90°C decontamination cycle and thermal conductivity (TC) CO2 sensor technology, 120V 50/60 Hz | 11676604 |
| 50153429 | Fisherbrand Isotemp CO2 incubator, stainless steel 150L chamber with on-demand 90°C decontamination cycle and infrared (IR) CO2 sensor technology, 120V 50/60 Hz | 11676605 |
| 50153430 | Fisherbrand Isotemp CO2 incubator, stainless steel 150L chamber with on-demand 90°C decontamination cycle and thermal conductivity (TC) CO2 sensor technology, 230V 50/60 Hz | 15660667 |
| 50153431 | Fisherbrand Isotemp CO2 incubator, stainless steel 150L chamber with on-demand 90°C decontamination cycle and infrared (IR) CO2 sensor technology, 230V 50/60 Hz | 15650667 |
Attachment(s):
| File | Last Modified |
|---|---|
| 50153099 - Fisherbrand C02 Incubators - Operations Manual.pdf | December 21, 2021 |