Thermo Fisher Scientific
7000310 - Rev 4 - Direct Heat CO2 Incubator, Forma Model 310 Series - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Direct Heat CO2 Incubator Forma Model 310 Series
Models:
Model CO2 Sensor* Voltage**
  • 310
T/C 115
  • 311
T/C 230
  • 320
IR 115
  • 321
IR 230
  • 350
T/C 115
  • 351
T/C 230
  • 360
IR 115
  • 361
IR 230
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60 HZ