7000370 - Steri-Cycle CO2 Incubator Direct Heat with Sterilization Cycle Model 370 Series - Operating and Maintenance Manual
The document "7000370 - Steri-Cycle CO2 Incubator, Direct Heat with Sterilization Cycle, Model 370 Series - Operating and Maintenance Manual" offers guidelines for operating and maintaining the Steri-Cycle CO2 Incubator, Direct Heat with Sterilization Cycle, for models 370, 371, 380, and 381. It details the types of sensors (thermal conductivity or infra-red) and voltage specifications (115V or 230V) for each model, covering the years 2001 to 2009.
Description:
Operating and maintenance manual, 2001 - 2009
Environment:
Steri-Cycle CO2 Incubator, Direct Heat with Sterilization Cycle, Model 370 Series:
| Model | Sensor* | Voltage** |
|
T/C | 115 |
|
T/C | 230 |
|
IR | 115 |
|
IR | 230 |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**All units are 50-60 Hz.