Thermo Fisher Scientific
7003010 - Water Jacketed CO2 Incubators, Model 3010 and 3020 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
  • Water Jacketed CO2 Incubators, model 3010
  • Water Jacketed CO2 Incubators, model 3020:
Model Sensor* Voltage**
  • 3010
T/C sensor 115V
  • 3020
IR sensor 115V
*TC is a thermal conductivity sensor, IR is an infra-red sensor.
**All units are 50-60Hz.