7003060 - Rev 1 - Portable IR CO2 Controller, Model 3060 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Portable IR CO2 Controller, model 3060, specifications:
Exterior Size | 8.000" wide x 3.000" high x 9.000" front to back |
Exterior Material | ABS Plastic, beige color |
Paint Slate Slate | Blue Insert, Powder Coated |
CO2 Range | 0 to 20% |
CO2 Connection | 1/4" Barbed Hose Fitting, 10-15 psi |
CO2 Sensor | 231238 Vaisala, 0.985" X 3.780", 4.5" leads leads |
CO2 Sensor Temperature Range | -20°C to +60°C |
CO2 Sensor Humidity Range | 0 to 100%, non-condensing |
Electrical | 100-250V, 1 PH, 2W, 50/60HZ, .7 FLA, Changeable AC plug |
Controller | 231235 Watlow PM Microprocessor |
Attachment(s):
File | Last Modified |
---|---|
7003060 - Rev 1 - Portable IR CO2 Controller, Model 3060 - Operating and Maintenance Manual.pdf | December 22, 2021 |