7003060 - Rev 1 - Portable IR CO2 Controller Model 3060 - Operating and Maintenance Manual
The purpose of the document "7003060 - Rev 1 - Portable IR CO2 Controller, Model 3060 - Operating and Maintenance Manual" is to provide detailed instructions for operating and maintaining the Portable IR CO2 Controller, model 3060. This includes specifications, usage guidelines, maintenance procedures, and troubleshooting information to ensure optimal performance and longevity of the device.
Description:
Operating and maintenance manual
Environment:
Portable IR CO2 Controller, model 3060, specifications:
| Exterior Size | 8.000" wide x 3.000" high x 9.000" front to back |
| Exterior Material | ABS Plastic, beige color |
| Paint Slate Slate | Blue Insert, Powder Coated |
| CO2 Range | 0 to 20% |
| CO2 Connection | 1/4" Barbed Hose Fitting, 10-15 psi |
| CO2 Sensor | 231238 Vaisala, 0.985" X 3.780", 4.5" leads leads |
| CO2 Sensor Temperature Range | -20°C to +60°C |
| CO2 Sensor Humidity Range | 0 to 100%, non-condensing |
| Electrical | 100-250V, 1 PH, 2W, 50/60HZ, .7 FLA, Changeable AC plug |
| Controller | 231235 Watlow PM Microprocessor |
Attachment(s):
| File | Last Modified |
|---|---|
| 7003060 - Rev 1 - Portable IR CO2 Controller, Model 3060 - Operating and Maintenance Manual.pdf | December 22, 2021 |