7003584 - NAPCO Series 8000DH CO2 Incubator Direct Heat with Sterilization Cycle - Operating and Maintenance Manual
The purpose of the "NAPCO Series 8000DH CO2 Incubator Direct Heat with Sterilization Cycle - Operating and Maintenance Manual" is to provide detailed instructions on the operation and maintenance of the NAPCO Series 8000DH CO2 Incubator. This manual includes information on how to use the incubator's direct heat and sterilization cycle features to ensure proper function and longevity of the equipment.
Description:
Operating and maintenance manual
Environment:
NAPCO Series 8000DH CO2 Incubator Direct Heat with Sterilization Cycle:
| Model | Sensor* | Voltage** | RHDisplay |
|
T/C | 115 | No |
|
IR | 115 | No |
|
T/C | 115 | Yes |
|
IR | 115 | Yes |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**All units are 50-60 Hz