Thermo Fisher Scientific
7010310 - Rev 20 - Forma Direct Heat CO2 Incubator, Model 310 Series - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Forma Direct Heat CO2 Incubator, Model 310 Series, models:
Model CO2 Sensor* Voltage** Temp °C Max
  • 310
T/C 115 55
  • 311
T/C 230 55
  • 320
IR 115 50
  • 321
IR 230 50
  • 350†
T/C 115 45
  • 351†
T/C 230 45
  • 360†
IR 115 45
  • 361†
IR 230 45
* TC is a thermal conductivity sensor. IR is an infrared sensor.
** All units are 50-60 Hz.
† Copper interior