Thermo Fisher Scientific
7013110 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Forma Model 3100, 3200 Series, models:
  • 3110 Series Single Chamber Water Jacket Incubator
  • 3210 Series Dual Chamber Water Jacket Incubator
Single Chamber Models
Model CO2 Sensor** O2 Voltage***
  • 3110
TC No 115
  • 3111
TC No 230
  • 3120
IR No 115
  • 3121
IR No 230
  • 3130
TC Yes 115
  • 3131
TC Yes 230
  • 3140
IR Yes 115
  • 3141
IR Yes 230
Dual Chamber Models
Model* CO2 Sensor** O2 Voltage***
  • 3210
TC No 115
  • 3211
TC No 230
  • 3220
IR No 115
  • 3221
IR No 230
  • 3230
TC Yes 115
  • 3231
TC Yes 230
  • 3240
IR Yes 115
  • 3241
IR Yes 230
*The models listed as dual chamber models are stacked or dual chamber units. These units arrive as single stand-alone units. They may be stacked one on top of the other or configured side by side with opposite door swing.
**TC is a thermal conductivity sensor. IR is an infrared sensor.
***All units are 50-60 HZ.