7013110 - Forma Model 3100_3200 Series - Operating and Maintenance Manual
The document titled "7013110 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual" provides comprehensive operating and maintenance instructions for the Forma Model 3100 and 3200 Series. This manual is essential for the proper use, maintenance, and troubleshooting of the 3110 Series Single Chamber Water Jacket Incubator and the 3210 Series Dual Chamber Water Jacket Incubator.
Description:
Operating and maintenance manual
Environment:
Forma Model 3100, 3200 Series, models:
- 3110 Series Single Chamber Water Jacket Incubator
- 3210 Series Dual Chamber Water Jacket Incubator
| Single Chamber Models | |||
| Model | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
| Dual Chamber Models | |||
| Model* | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*The models listed as dual chamber models are stacked or dual chamber units. These units arrive as single stand-alone units. They may be stacked one on top of the other or configured side by side with opposite door swing.
**TC is a thermal conductivity sensor. IR is an infrared sensor.
***All units are 50-60 HZ.
Attachment(s):
| File | Last Modified |
|---|---|
| 7013110 - Rev 0 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual.pdf | January 25, 2022 |
| 7013110 - Rev 1 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual.pdf | January 25, 2022 |