7023110 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Forma Model 3100/3200 Series, models:
- 3110 Series Single Chamber Water Jacket Incubator
- 3210 Series Dual Chamber Water Jacket Incubator
Single Chamber Models | |||
Model | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
Dual Chamber Models | |||
Model* | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*The models listed as dual chamber models are stacked or dual chamber units. These units arrive as single stand-alone units. They may be stacked one on top of the other or configured side by side with opposite door swing.
**TC is a thermal conductivity sensor. IR is an infrared sensor.
***All units are 50-60 HZ.