Thermo Fisher Scientific
7043110 - Forma Series II Water Jacket CO2 Incubator, Model 3100 Series - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Forma Series II, Water Jacket CO2 Incubator, Model 3100 Series
Single chamber models:
Model CO2 Sensor* O2 Voltage **
  • 3110
TC No 115
  • 3111
TC No 230
  • 3120
IR No 115
  • 3121
IR NO 230
  • 3130
TC Yes 115
  • 3131
TC Yes 230
  • 3140
IR Yes 115
  • 3141
IR Yes 230
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60 Hz.