7043110 - Forma Series II Water Jacket CO2 Incubator Model 3100 Series - Operating and Maintenance Manual
The document titled "7043110 - Forma Series II Water Jacket CO2 Incubator, Model 3100 Series - Operating and Maintenance Manual" provides comprehensive operating and maintenance instructions for the Forma Series II Water Jacket CO2 Incubator, Model 3100 Series. This manual is essential for the proper use, maintenance, and troubleshooting of single chamber models in this series.
Description:
Operating and maintenance manual
Environment:
Forma Series II, Water Jacket CO2 Incubator, Model 3100 Series
Single chamber models:
| Model | CO2 Sensor* | O2 | Voltage ** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | NO | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60 Hz.