Thermo Fisher Scientific
7053110 - Forma Series II Water Jacket CO2 Incubator, Model 3100 Series - Operating and Maintenance Manual
Description: 
 Operating and maintenance manual
Environment:
Forma Series II Water Jacket CO2 Incubator, model 3100 Series:
Model CO2 Sensor* O2 Voltage ** Temp Max
  • 3110
TC No 115 55C
  • 3111
TC No 230 55C
  • 3120
IR No 115 50C
  • 3121
IR No 230 50C
  • 3130
TC Yes 115 45C
  • 3131
TC Yes 230 45C
  • 3140
IR Yes 115 45C
  • 3141
IR Yes 230 45C
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60 Hz