7053110 - Forma Series II Water Jacket CO2 Incubator, Model 3100 Series - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Forma Series II Water Jacket CO2 Incubator, model 3100 Series:
Model | CO2 Sensor* | O2 | Voltage ** | Temp Max |
|
TC | No | 115 | 55C |
|
TC | No | 230 | 55C |
|
IR | No | 115 | 50C |
|
IR | No | 230 | 50C |
|
TC | Yes | 115 | 45C |
|
TC | Yes | 230 | 45C |
|
IR | Yes | 115 | 45C |
|
IR | Yes | 230 | 45C |
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60 Hz