Thermo Fisher Scientific
88732 - Rev E - Fisher Isotemp Low Temperature Incubator, Model 307C - User Manual and Parts List
Description: 
User manual and parts list
Environment:
Fisher Isotemp Low Temperature Incubator, model 307C and catalog number 11-679-25C:
Power Requirements: 115V, 8.9A, 60 Hz Including 2 A for convenience outlet
Volume: 20.3 cu ft 0.58m3