LT1448X1 - Air-Jacketed Compact Flo-Thru CO2 Incubators - Operating Manual and Parts List
The Thermo Scientific Air-Jacketed Compact Flo-Thru CO2 Incubators Operating Manual and Parts List provides detailed operational instructions and a comprehensive parts list for the Air-Jacketed Compact Flo-Thru CO2 Incubator models 315 and 315DV. These models require either 120 volts at 200 watts and 1.7 amps or 240 volts at 200 watts and 0.8 amps, operating at 50/60 Hz.
Description:
Operating manual and parts list
Environment:
Thermo Scientific Air-Jacketed Compact Flo-Thru CO2 Incubator:
| Model No. | Power Requirements |
|
120 Volts, 50/60 Hz, 200 Watts, 1.7 Amps |
|
240 Volts, 50/60 Hz, 200 Watts, 0.8 Amps |
Attachment(s):
| File | Last Modified |
|---|---|
| LT1448X1 - Air-Jacketed Compact Flo-Thru CO2 Incubators - Operating Manual and Parts List.pdf | December 23, 2021 |