Thermo Fisher Scientific
LT1465X3 - Imperial III General Purpose Incubators, Series 1464, 1465, 1466 - User Manual
Description: 
User manual
Environment:
Barnstead Lab-Line Imperial III General Purpose Incubators, Series 1464, 1465, and1466 
Model numbers and electrical requirements:
Model No. Voltage Hertz Amps Watts  
  • 302
  • 120
  • 50-60
  • 2.1
  • 400
  • 302-1
  • 240
  • 50-60
  • 0.7
  • 400
  • 305
  • 120
  • 50-60
  • 3.3
  • 400
  • 1 electrical outlet, 120V, 500W total load
  • 305M
  • 120
  • 50-60
  • 3.7
  • 400
  • 1 electrical outlet, 120V, 500W total load
  • 305-1
  • 240
  • 50-60
  • 1.7
  • 400
  • 1 electrical outlet, 120V, 500W total load
  • 305M-1
  • 240
  • 50-60
  • 1.7
  • 400
  • 1 electrical outlet, 120V, 500W total load
  • 310
  • 120
  • 50-60
  • 5.0
  • 600
  • 1 electrical outlet, 120V, 500W total load
  • 310M
  • 120
  • 50-60
  • 5.8
  • 600
  • 1 electrical outlet, 120V, 500W total load
  • 310-1
  • 230-240
  • 50-60
  • 2.5
  • 600
  • 1 electrical outlet, 120V, 500W total load
  • 310M-1
  • 230-240
  • 50-60
  • 2.5
  • 600
  • 1 electrical outlet, 120V, 500W total load