50164798-a - CW3 Modified Saline Sensor Kit - Instructions
To connect a saline cube to a CW3 Cell Washer using the modified saline sensor kit (75007410), follow the steps in this article: 1. Place the saline cube at a position not higher than the cell washer, with the opening facing upwards. 2. Insert the liquid sensor and the inlet tube into the saline cube. 3. Loosen the cord lock and tighten the screw cap onto the opening of the saline cube. 4. Adjust the liquid sensor to the bottom of the saline cube and secure it...
Description:
Instructions for sensor kit
Environment:
CW3, CW-3, 75007410,
Attachment(s):
| File | Last Modified |
|---|---|
| 50164798_a_Modified Saline Sensor Kit-multi.pdf | November 04, 2022 |