Sorvall ST1 Plus Series and Legend X1 Pro Series - Thermo Scientific SL1 Plus _ Multifuge X1 Pro Series_Megafuge ST1 Plus - Emergency Release Instructions Update
Issue
NEW GP 1 Pros and Plus Series shows the manual lid release cord in the back of the unit not on the left side.
Environment
Sorvall Legend X1 Pro
Sorvall Legend X1R Pro
Sorvall ST1 Plus
Sorvall ST1R Plus
Thermo Scientific SL1 Plus
Thermo Scientific SL1R Plus
Thermo Scientific Multifuge X1 Pro
Thermo Scientific Multifuge X1R Pro
Thermo Scientific Megafuge ST1 Plus
Thermo Scientific Megafuge ST1R Plus
Resolution
In the event of a power failure, the centrifuge lid cannot be opened using the regular electric lid release. A mechanical override is available for emergency sample recovery. This should only be used in emergencies and after the rotor has completely stopped. Please follow these steps carefully:
-
Ensure the Rotor Has Stopped: Look through the view port in the lid to confirm that the rotor has come to a complete stop. Wait a few minutes as the braking process takes longer without power.
-
Disconnect the Power Supply: Unplug the power supply to ensure safety.
-
Access the Mechanical Release:
- Locate the white plastic plug on the left side of the housing.
- Use a screwdriver or knife to pry out the plug.
- Pull the release cord attached to the plug to trigger the mechanical lid release. The lid will open, allowing you to remove the samples.
-
Secure the Release Cord: Push the release cord back into the centrifuge and reinsert the plug.
-
Restore Power and Reactivate Door Locks: Once power is restored, reconnect the centrifuge. Switch it on and press the OPEN key to reactivate the door locks.
Attachment(s)
| File | Last Modified |
|---|---|
| GP1 Pros Release Pic.jfif | July 17, 2025 |