Technical Data Sheet - Sorvall BP8_BP16_BIOS 16 - Thermo Scientific Cryofuge 8 and Cryofuge 16
The Thermo Scientific Cryofuge 8 and Cryofuge 16 technical data sheet details specifications for these centrifuges. Key features include maximum speeds of 5000 rpm for Cryofuge 8 and 4700 rpm for Cryofuge 16, with RCF values of 7295 x g and 7187 x g, respectively. They operate within a temperature range of -20°C to 40°C and have noise levels of 62 dB(A). These centrifuges are compatible with multiple voltage configurations and comply with international standards, including FDA, CE, and RoHS.
Description:
Technical Data Sheet
Environment:
Sorvall BP8
Sorvall BP16
Sorvall BIOS 16
Heraeus Cryofuge 16
Heraeus Cryofuge 8
Attachment(s):
| File | Last Modified |
|---|---|
| Technical Data Sheet - BP8 BP16 BIOS16 Cryofuge 8 Cryofuge 16.pdf | May 27, 2022 |