Technical Data Sheet - Sorvall BP8 HD_BP16 HD_BIOS 16 HD_BIOS A HD - Thermo Scientific Cryofuge 16 HD
The technical data sheet for Sorvall BP8 HD, BP16 HD, BIOS 16 HD, BIOS A HD, and Thermo Scientific Cryofuge 16 HD provides specifications on environmental conditions, electrical requirements, performance, rotor specifications, dimensions, weight, noise levels, and temperature settings. These centrifuges are designed for indoor use, with specific voltage and power outputs, and have varying rotor capacities and maximum speeds. Noise levels at maximum speed are 62 dB, and temp set -20°C to +40C
Description:
Technical Data Sheet
Environment:
BP8 HD
BP16 HD
BIOS 16 HD
BIOS A HD
Cryofuge 16 HD
Attachment(s):
| File | Last Modified |
|---|---|
| Technical Data Sheet - BP8HD, BP16HD, BIOS16HD.pdf | May 27, 2022 |