45912 Rev 2 - Sorvall Discovery 90 SE and Discovery 100 SE - User Manual
The DiscoveryTM 100 SE and DiscoveryTM 90 SE Ultracentrifuges are designed and manufactured based on our long experience in the development of centrifuges; they are easy to use and highly reliable. Features include the following: 1. Maximum speed of 100,000 rpm (802,006 x g) 2. Control Panel is simple with easy key operation and easy-to-see liquid crystal screen. 3. The displayed language can be switched over between English and Japanese. 4. The real-time control (RTC) feature enables setting a
Description:
Operating instructions
Environment:
- Sorvall Discovery 100 SE Ultracentrifuge
- Sorvall Discovery 90 SE Ultracentrifuge
Attachment(s):
| File | Last Modified |
|---|---|
| 45912-2 - Sorvall Discovery 90, 100 SE - User Manual | January 28, 2022 |