50145792 Rev B - Thermo Scientific Sorvall WX Plus Series Ultracentrifuges - Instruction Manual
The Thermo Scientific Sorvall WX Plus Series Ultracentrifuges Instruction Manual provides comprehensive guidance on the operation, features, and specifications of these ultracentrifuges. Key features include a maximum speed of 100,000 rpm, an integrated touchscreen interface, multiple language options, real-time control, and programmable modes. The manual also highlights the space-saving design, quiet operation, and various alert indicators for troubleshooting.
Description:
Instruction manual
Environment:
Thermo Scientific Sorvall WX+ Ultracentrifuge
WX-90+
WX-80+
WX-100+
WX-80 Plus
WX-90 Plus
WX-100 Plus
WX 80+
WX 90+
WX 100+
WX 80 Plus
WX 90 Plus
WX 100 Plus
Attachment(s):
| File | Last Modified |
|---|---|
| 50145792-b - Thermo Scientific Sorvall WX+ Ultracentrifuge - Instruction Manual.pdf | December 07, 2021 |