Thermo Fisher Scientific
56311 Rev 6 - Sorvall RC3BP - Operating Instructions
The Sorvall RC3BP Operating Instructions provide comprehensive guidance on the operation and maintenance of the Sorvall RC3BP Lowspeed Refrigerated Centrifuge. Key sections include installation, detailing inspection and electrical requirements, and operation, covering power-up procedures, rotor installation, loading, and balancing. The centrifuge, designed for regulated environments like blood banks, operates up to 6000 rpm, generating forces up to 7277g.
Description: 
This manual contains information required to operate and maintain the SORVALL® RC3BP™ Lowspeed Refrigerated Centrifuge.
Environment:
Sorvall RC3BP
RC-3BP
RC 3BP
  • Was this article helpful?