Thermo Fisher Scientific
S999879P0 - Sorvall CC40 Large-Scale Continuous Flow Centrifuge - Instruction Manual
The Sorvall CC40 Large-Scale Continuous Flow Centrifuge Instruction Manual provides comprehensive guidance on operating the centrifuge, including its features, safety protocols, and troubleshooting. The centrifuge achieves a maximum speed of 40,000 rpm and a centrifugal force of 118,000 x g. It features a high-frequency motor drive system, a digital control system for monitoring run conditions, and a failure self-diagnosis function for easy troubleshooting.
Description: 
Instruction manual
Environment:
SORVALL CC40 Large-Scale Continuous Flow Centrifuge
(USP class O-ring)
  • Was this article helpful?