Sorvall BP8_BP16_BIOS 16 Low Speed Centrifuge - Installation Checklist
The installation checklist for the Sorvall BP8, BP16, and BIOS 16 Low Speed Centrifuge includes verifying account information, contacting the customer to confirm receipt of the instrument and accessories, and ensuring space and proper electrical setup. On-site inspection covers shipping damage and parts inventory. Installation involves checking voltage, grounding, and leveling. A test run ensures functionality, followed by customer training on operation and maintenance.
Description:
Installation checklist
Environment:
- SORVALL Centrifuges BP8 low Speed Centrifuge
- SORVALL Centrifuges BP16 low Speed Centrifuge
- SORVALL Centrifuges BIOS16 low Speed Centrifuge
Attachment(s):
| File | Last Modified |
|---|---|
| SORVALL Centrifuges BP8 BP16 BIOS 16 Low Speed Centrifuge - Installation Checklist.pdf | December 08, 2021 |