Sorvall Lynx 6000 Centrifuge - Pre-Installation Checklist
The pre-installation checklist for the Sorvall Lynx 6000 Centrifuge includes requirements for power receptacle and source, ambient temperature, location, physical dimensions, and ventilation. Ensure a dedicated 30 amp power source with proper voltage, maintain ambient temperatures within specified range, install on a flat, vibration-free surface, and provide adequate ventilation. The centrifuge dimensions are 27.6 inches wide, 31.7 inches deep, and 36.6 inches high.
Description:
Pre-installation checklist
Environment:
SORVALL Lynx 6000 Centrifuge
Lynx 6000
Attachment(s):
| File | Last Modified |
|---|---|
| SORVALL Lynx 6000 Centrifuge - Pre-Installation Checklist.pdf | December 08, 2021 |