Heraeus Megafuge 16R Acceleration and Deceleration Curves
The Heraeus Megafuge 16R centrifuge features detailed acceleration and deceleration profiles for various rotors and buckets. For example, the HighConic II 6x100 rotor accelerates from 0 to 9000 rpm in approximately 4 minutes and 30 seconds, and decelerates back to 0 in around 28 minutes. The BIOShield 720 rotor reaches 5500 rpm in the same acceleration time, with deceleration taking about 26 minutes. These profiles aid in understanding the centrifuge's performance with different setups
Description:
Acceleration and deceleration curves
Environment:
Heraeus Megafuge 16R
Attachment(s):
| File | Last Modified |
|---|---|
| Heraeus Megafuge 16R 120V Acceleration, Deceleration Curves.pdf | December 08, 2021 |
| Heraeus Megafuge 16R 230V Acceleration, Deceleration Curves.pdf | December 08, 2021 |