Heraeus Megafuge 16 Acceleration and Deceleration Curves
The Heraeus Megafuge 16 features distinct acceleration and deceleration curves for different rotors. For example, the HighConic II 6x100 rotor accelerates to 9000 rpm in about 4 minutes 30 seconds and decelerates to 0 over 28 minutes. Similarly, the BIOShield 720 rotor reaches 5500 rpm in the same acceleration time and decelerates in 26 minutes. Profiles for other rotors like FA 30x15 and Microplate M-20 detail varying speeds and times, essential for optimizing centrifuge operations.
Description:
Acceleration and deceleration curves
Environment:
Heraeus Megafuge 16
Attachment(s):
| File | Last Modified |
|---|---|
| Heraeus Megafuge 16 230V Acceleration, Deceleration Curves.pdf | December 08, 2021 |
| Heraeus Megafuge 16 Acceleration, Deceleration Curves.pdf | December 08, 2021 |