Heraeus Multifuge X1R Acceleration Deceleration Curves
The Heraeus Multifuge X1R features detailed acceleration and deceleration curves for various rotors and buckets. For example, the Microliter 48x2 sealed rotor accelerates from 0 to 16000 rpm in up to 5 minutes and decelerates in up to 34 minutes. The Highconic II 6x100 rotor accelerates to 11000 rpm in 4.5 minutes and decelerates in 30 minutes. These profiles ensure optimized performance and safety for different applications.
Description:
Acceleration and deceleration curves
Environment:
Heraeus Multifuge X1R
Attachment(s):
| File | Last Modified |
|---|---|
| Heraeus Multifuge X1R 120V Acceleration, Deceleration Curves.pdf | December 08, 2021 |
| Heraeus Multifuge X1R 230V Acceleration, Deceleration Curves.pdf | December 08, 2021 |