Fisher Scientific AccuSpin 8c - Acceleration and Deceleration Curves
The Fisher Scientific accuSpin 8C Clinical Centrifuge features one standard acceleration profile and three deceleration profiles: standard, soft, and brake-off. The deceleration profile can be adjusted using the Curve Key, with settings indicated by LEDs. These profiles are based on a fully loaded rotor at rated voltage and can vary with conditions. Diagrams for fixed angle and swinging bucket setups illustrate time-speed relationships for each profile, providing reference for operation.
Description:
Acceleration and deceleration curves
Environment:
Fisher Scientific AccuSpin 8C
Attachment(s):
| File | Last Modified |
|---|---|
| Fisher Scientific AccuSpin 8C Acceleration, Deceleration Curves.pdf | December 08, 2021 |