Thermo Scientific SL16 Acceleration and Deceleration Curves
The Thermo Scientific SL16 centrifuge features detailed acceleration and deceleration profiles for various rotors. For example, the Highconic II 6x100 rotor accelerates from 0 to 9000 rpm in up to 4:30 minutes and decelerates from 9000 to 0 rpm in up to 28:00 minutes. Other rotors, like the FA Rotor 30x15 and Microplate Rotor M-20, have specific profiles tailored to their maximum speeds and durations.
Description:
Acceleration and deceleration curves
Environment:
Thermo Scientific SL16
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific SL16 230V Acceleration, Deceleration Curves.pdf | December 08, 2021 |
| Thermo Scientific SL16 120V Acceleration, Deceleration Curves.pdf | December 08, 2021 |