11243-22 - Thermo Fisher Scientific H-6000A and HBB-6 and HLR-6 Rotor - Instruction Manual
This manual contains information required to operate and maintain the Thermo Scientific H-6000A, HBB-6, or HLR-6 Swinging Bucket Rotor. If you require additional information regarding operation or maintenance, please contact Thermo Fisher Scientific for assistance. The Thermo Scientific H-6000A, HBB-6, and HLR-6 are six-place swinging bucket rotors designed for use in the Thermo Scientific RC 3B, RC 3B+, RC 3C, RC 3C+, RC 3BP™ and RC 3BP™+ Centrifuges. Each rotor has a maximum speed of 5000
Description:
This manual is a guide for the use of Thermo Scientific H-6000A, HBB-6 and HLR-6 rotors
Environment:
The Thermo Scientific
- H-6000A
- HBB-6
- HLR-6
are six-place swinging bucket rotors designed for use in the Thermo Scientific:
- RC 3B Centrifuges
- RC 3B+ Centrifuges
- RC 3C Centrifuges
- RC 3C+ Centrifuges
- RC 3BP Centrifuges
- RC 3BP+ Centrifuges
Attachment(s):
| File | Last Modified |
|---|---|
| 11243-22 - Thermo Fisher Scientific H-6000A, HBB-6, and HLR-6 Rotor - Instruction Manual.pdf | December 09, 2021 |