79369 Rev 3 - Sorvall Surespin 630 Swinging Bucket Ultracentrifuge Rotor - Operating Instructions
The Sorvall Surespin 630 Swinging Bucket Ultracentrifuge Rotor is a titanium rotor designed to process up to six tubes at speeds up to 30,000 rpm. The article details the rotor's description, specifications, and operating instructions, emphasizing safety checks before each run. Key points include ensuring clean rotor components, proper installation of bucket caps, and verifying chemical compatibility. The rotor's buckets use a precision thread design for secure sealing.
Description:
Operating instructions
Environment:
SORVALL Surespin 630 Titanium Swinging Bucket Ultracentrifuge Rotor
Attachment(s):
| File | Last Modified |
|---|---|
| 79369-3 - SORVALL Surespin 630 Swinging Bucket Ultracentrifuge Rotor - Operating Instructions.pdf | December 09, 2021 |