Ultracentrifuge Rotors - Tubes Bottles and Adapters Rotor Guide and Specifications
The Thermo Scientific Ultracentrifuge Rotors guide provides detailed specifications for various rotors, tubes, bottles, and adapters. Key rotors include the TN-890 with a capacity of 8 x 5 mL, max speed of 90,000 rpm, and the TH-660 with a capacity of 6 x 4.4 mL, max speed of 60,000 rpm. Each rotor package includes necessary accessories like caps, stands, and manuals. Tubes and bottles are specifically designed for each rotor, ensuring optimal performance and compatibility.
Description:
Rotor guide and specifications
Environment:
Thermo Scientific Ultracentrifuge Rotors:
| Fixed Angle: |
|
|
|
|
|
|
|
|
|
|
|
|
|
|
| Swinging Bucket: |
|
|
|
|
|
|
|
| Vertical: |
|
|
|
|
|
| Neo Angle: |
|
|
|
| Zonal: |
|
|
| Continuous Flow: |
|
Attachment(s):
| File | Last Modified |
|---|---|
| DSCFGULTRATZ28 1014 - Thermo Scientific Ultracentrifuge Rotors - Rotor Guide and Specifications.pdf | January 06, 2022 |