GP1 and GP3 - Error List for Troubleshooting
The error list for troubleshooting GP1 and GP3 centrifuges includes various error codes and their descriptions. Common issues include unexplained imbalances (E-20, E-21, E-22, E-44, E-98), operational errors (E-44, E-45), and main PCB errors (E-97). Detailed troubleshooting steps involve checking mechanical issues, ensuring proper loading, and addressing PCB problems.
Description:
Error description for GP-Centrifuge family
Environment:
- GP1 Series
- GP3 Series
Attachment(s):
| File | Last Modified |
|---|---|
| GP1 and GP4 - Error List for Troubleshooting | January 14, 2022 |